,,19 components: 100mg/l each of Ce Dy Er Eu Ga Gd Ho In La Lu Nd Pr Sm Tb Th Tm U Y Yb in HNO3 2% ,,CRM, ISO 17034 and ISO 17025,,,, ,,Conservation 24 months.,,,, ,,Store under normal laboratory conditions, at tem 1 * 100 mL
Catalogus # CMPLRM109681L1
Leverancier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together