[EN]BIOTAINER 1.8L 4GU NEW KIT COMPLET 1 * 24 ST
Catalog # CORT3BIO184GU
Supplier: E3 Cortex
New Product
[EN]BIOTAINER 1.8L 4GU NEW KIT COMPLET 1 * 24 ST
Catalog # CORT3BIO184GU
Supplier: E3 Cortex
New Product
Frequently Bought Together
Cleanroom handschoenen, nitril, BioClean Nerva™
Supplier: Nitritex
Nitril handschoenen, TouchNTuff® 93-250
Supplier: Ansell
Nitril handschoenen, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Laboratoriummarkers
Supplier: VWR Collection
VWR®, Laboratoriumjassen, Howie
Supplier: VWR Collection
Laboratory coats, C851
Supplier: Portwest
GHS-pictogrammen
Supplier: Brady
Latex handschoenen, TouchNTuff® 69-318
Supplier: Ansell
VWR®, Nitrilhandschoenen
Supplier: VWR Collection
Industrial gloves, AlphaTec® 58-735
Supplier: Ansell
VWR® Basic Cleanroom-overall, SMS
Supplier: VWR Collection
Colour Lab Tapes
Supplier: GA
Snijbestendige handschoenen, ActivArmr® 80-658
Supplier: Ansell
Reinigingsgereedschap, EasyReach™
Supplier: CONTEC
High temperature anti-static cleanroom tapes
Supplier: ULTRATAPE
Hi-visibility Warsaw executive waistcoats, Vest-Port, C476
Supplier: Portwest
ESD safety shoes, BRICE XXTM, low, S3
Supplier: ELTEN