Orders LinkContactUsLinkComponent
Corning® 25-Layer CellCube® Module, Corning
Corning® 25-Layer CellCube® Module, Corning
Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"
Catalogus # CORN3241
Leverancier:   Corning
Corning® 25-Layer CellCube® Module, Corning
Catalogus # CORN3241
Leverancier:  Corning
Corning® 25-Layer CellCube® Module, Corning
Corning® 25-Layer CellCube® Module, Corning
Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"
Price
913,00 €
Log in voor beschikbaarheid
1 * 5 ST
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Documents
Specificaties
  • Omschrijving:
    Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"
  • Seq:
    03