Accessories for Continuous Level Control, SymLine®
Built-in Signalbox1, EU/UK/US, 1-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for one container, one switch input and one switch output for peripheral units, including 24 V power supply
Catalogus # SCTV106741
Leverancier: S.C.A.T.
Ga naar:
Product Details & Documents
LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.
- Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
- Touchless alarm-resetting fields for two containers
- Two switch inputs and two switch outputs for peripheral units
- Incl. 230 V power supply cable with open end and clamp
Suitable e.g. for equipment from Wesemann, Waldner, Bense, Hemling, Köttermann, Kewaunee, Carlo Erba, Modus Complete, Delagrave, and many more.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specificaties
- Omschrijving:Built-in Signalbox1, EU/UK/US, 1-channel, for safety waste cap and other electronical level controlers, touchless alarm-resetting fields for one container, one switch input and one switch output for peripheral units, including 24 V power supply
- Seq:5