Orders LinkContactUsLinkComponent
Toebehoren
Toebehoren
Polarizer slider A, 6×30 mm, 90° rotatable usable with reflected-light illuminator HD DIC for microscope Axiolab 5 MAT model
Catalogus # ZEIS4277109000000
Leverancier:   ZEISS
Toebehoren
Catalogus # ZEIS4277109000000
Leverancier:  ZEISS
Toebehoren
Toebehoren
Polarizer slider A, 6×30 mm, 90° rotatable usable with reflected-light illuminator HD DIC for microscope Axiolab 5 MAT model
Price
815,07 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Polarizer slider A, 6×30 mm, 90° rotatable usable with reflected-light illuminator HD DIC for microscope Axiolab 5 MAT model
  • Seq:
    09