[EN]SYRINGE 2 PART LUER LOCK 50ML 1 * 30 ST
Catalogus # AIRTAML50
Leverancier: Air-Tite Products
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
VWR®, gezichtstissues
Leverancier: VWR Collection
VWR®, Nitrilhandschoenen
Leverancier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Flessen, smalle hals, rond, HDPE, met schroefdoppen, Nalgene®
Leverancier: Thermo Fisher Scientific
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Teststrips, peroxidetest, MQuant™, Supelco®
Leverancier: Merck
Cell culture storage bottles, polystyrene, Corning®
Leverancier: Corning
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
Leverancier: VWR Collection
Gel filtration chromatography columns, standard, CHROMAFLEX®
Leverancier: DWK Life Sciences
Chemisch bestendige handschoenen, Butoject® 898
Leverancier: KCL by Honeywell