Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Genistein

Glass bottle for solids, 1 g
Catalogus # ENZOALX350006G001
Leverancier:   Enzo Life Sciences
Genistein
Catalogus # ENZOALX350006G001
Leverancier:  Enzo Life Sciences
CAS-nummer:  446-72-0
Genistein

Glass bottle for solids, 1 g

CAS-nummer:  446-72-0
Price
1 030,00 €
Log in voor beschikbaarheid
1 * 1 g
Product Details & Documents
Synoniemen:  5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
  • CAS-nummer:446-72-0
  • Formule:C₁₅H₁₀O₅
  • MDL-nummer:MFCD00016952
  • Molecular Weight:270.24 g/mol
  • Smelten Pt:297…298 °C
  • EINECS:207-174-9
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    1 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.