Pressure Transducers
Pressure transducer, 1×1" TC, polysulfone, non sterile, −10 - +75 psi
Catalogus # SPECACPM-10TC-01N
Leverancier: Spectrum Laboratories
Ga naar:
Product Details & Documents
Specificaties
- Meetbereik:−10 - +75 psi
- Omschrijving:Pressure transducer, 1×1" TC, polysulfone, non sterile
- Seq:04
- Cat. No.:SPECACPM-10TC-01N
- Nauwkeurigheid:±2% from 0 - 6 psi, ±3% from 6 - 30 psi