Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Bottles, Square, HDPE

Bottles, Square, HDPE

100 ml, OV 32, HDPE, wit, 45×45×101 mm
Catalogus # RIXI2-0201-0100-32
Bottles, Square, HDPE
Catalogus # RIXI2-0201-0100-32
Bottles, Square, HDPE

100 ml, OV 32, HDPE, wit, 45×45×101 mm

Price
40,40 €
Log in voor beschikbaarheid
1 * 20 ST
Product Details & Documents

Square bottles made up of HDPE plastic, white.

Specificaties
  • Schroefdraad:
    OV 32
  • Capaciteit:
    100 ml
  • Kleur:
    wit
  • Materiaal:
    HDPE
  • Seq:
    01
  • b×d×h:
    45×45×101 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.