Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
coelenterazine ≥95%

coelenterazine ≥95%

Plastic bottle, 10 mg
Catalogus # AATB21166
Irriterend
Leverancier:   AAT Bioquest
coelenterazine ≥95%
Catalogus # AATB21166
Leverancier:  AAT Bioquest
CAS-nummer:  55779-48-1
coelenterazine ≥95%
coelenterazine ≥95%

Plastic bottle, 10 mg

CAS-nummer:  55779-48-1
Price
785,00 €
Log in voor beschikbaarheid
1 * 10 mg
Product Details & Documents

Calcium measurement is critical for numerous biological investigations.

Synoniemen:  8-Benzyl-2-(4-hydroxybenzyl)-6-(4-hydroxyphenyl)imidazo[1,2-a]pyrazin-3(7H)-one, CLZN, 2-[(4-Hydroxyphenyl)methyl]-6-(4-hydroxyphenyl)-8-(phenylmethyl)-imidazo [1,2-a] pyrazin-3-(7H)-one
  • CAS-nummer:55779-48-1
  • Formule:C₂₆H₂₁N₃O₃
  • MDL-nummer:MFCD00467176
  • Molecular Weight:423.472 g/mol
  • ADR:5,II
Specificaties

Specificaties

  • Verpakking:
    Plastic bottle
  • VPE:
    10 mg

Specificaties