Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
coelenterazine ≥95%

coelenterazine ≥95%

Plastic bottle, 250 µG
Catalog # AATB21150
Irritant
Supplier:   AAT Bioquest
coelenterazine ≥95%
Catalog # AATB21150
Supplier:  AAT Bioquest
CAS Number:  55779-48-1
coelenterazine ≥95%
coelenterazine ≥95%

Plastic bottle, 250 µG

CAS Number:  55779-48-1
Price
122,00 €
Log in for availability
1 * 250 µG
Product Details & Documents

Calcium measurement is critical for numerous biological investigations.

Synonyms:  8-Benzyl-2-(4-hydroxybenzyl)-6-(4-hydroxyphenyl)imidazo[1,2-a]pyrazin-3(7H)-one, CLZN, 2-[(4-Hydroxyphenyl)methyl]-6-(4-hydroxyphenyl)-8-(phenylmethyl)-imidazo [1,2-a] pyrazin-3-(7H)-one
  • CAS Number:55779-48-1
  • Formula:C₂₆H₂₁N₃O₃
  • MDL Number:MFCD00467176
  • Molecular Weight:423.472 g/mol
  • ADR:5,II
Specifications

Specifications

  • Verpakking:
    Plastic bottle
  • VPE:
    250 µG

Specifications