Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Cleanroom Socks

Cleanroom Socks

wit, L/XL
Catalogus # ISOF70610L-XL
Leverancier:   ISOFIELD IRELAND
Cleanroom Socks
Catalogus # ISOF70610L-XL
Leverancier:  ISOFIELD IRELAND
Cleanroom Socks
Cleanroom Socks

wit, L/XL

Price
652,00 €
Log in voor beschikbaarheid
1 * 500 PAIRE
Product Details & Documents

Non-sterile, lightweight, single-use cleanroom socks.

  • 95% polyester, 5% spandex
  • Mid-calf length
  • White colour
  • Three sizes for added flexibility
  • Low linting

Store in a dry, cool place (<40 °C) away from direct sunlight. Do not expose open cartons to prolonged direct fluorescent light.

Cleanroom socks are designed for single-use application in pharmaceutical product preparation, cleanroom cleaning and preparation, medical device manufacturing, blending operations for biotechnologies, handling of active materials, and maintenance of equipment and instruments.

Shelf life: Five years from date of manufacture.

Certificeringen: ISO Class 4 and 5 cleanroom, Class 10 and 100 cleanroom. Manufactured in a facility operating under ISO 9001:2015 quality management system. Processed in a NEBB certified ISO Class 5 cleanroom.

Verpakking: 10 pairs per double bag, 50 sealed outer cleanroom bags per carton, 500 pairs per carton.

Documents

Specificaties
  • Confectiemaat:
    L/XL
  • Kleur:
    wit
  • Cat. No.:
    ISOF70610L-XL
  • Kledingstuk toepassing:
    Pharmaceutical|Laboratory
  • Cleanroomklasse:
    10/ISO 4
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.