[EN]FOAM SWAB ANTI-STATIC 1 * 50 ST
CTSACTS-21936
: SCIENCIX
Frequently Bought Together
Spare Parts for VWR® Hitachi HPLC Instruments
: VWR Collection
[EN]PELTIER SENSOR 1 CORD 1 * 1 ST
: VWR Collection
BATTERIJ 3V VOOR MULTIPETTE PLUS CR2032 1 * 1 ST
: EPPENDORF
Accessories for Pipet-Aid® controllers
: DRUMMOND
[EN]START-UP SVC (BSC W/O PREFIL.) 1 * 1 ST
: Thermo Scientific
[EN]REPLACEMENT FEED TUBING PTFE CUT 300 1 * 1 ST
: Socorex Isba
SILICONEN VET VR FINNPIPETTE 1 * 100 g
: Thermo Fisher Scientific
SERVICE KIT 2ND LEVEL, P200 1 * 1 ST
: GILSON
[EN]STAND 500 MM STAINLESS STEEL 1 * 1 ST
: HIRSCHMANN
Various replacement parts
: SCIENCIX