MagicPrep™ NGS System
Product Details & Documents
MagicPrep™ NGS system is a new category of automated laboratory equipment that provides push button simplicity for NGS library preparation. The Revelo DNA-Seq Enz for MagicPrep™ NGS library preparation kit provides all the reagents for generating DNA-Seq libraries on the MagicPrep™ NGS system.
- Simple run setup in 10 minutes
- Reliable and reproducible
- True walk-away automation
- Pre-optimised DNA fragmentation for a broad input range simplifies the workflow
- Provides even coverage and uniformity across the bacterial genome regardless of GC content
- High quality libraries from pre-fragmented DNA
- Fully automated workflow
- Start with total RNA and MagicPrep™ NGS does all the library preparation steps, including selection of polyadenylated transcripts
- Even coverage across the entire transcript enabling detection of splice variants and isoforms during data analysis
- High quality data with consistent gene detection across a 100-fold difference in input
MagicPrep™ NGS system is a new category of automated laboratory equipment that provides push button simplicity for NGS library preparation for Illumina sequencing platforms.The MagicPrep™ NGS system provides a complete solution that includes the instrument, software, pre-optimized scripts, reagents and consumables.The MagicPrep™ NGS system integrates everything you need for library preparation; all you need to do is add your samples.
The Revelo DNA-Seq Enz for MagicPrep™ NGS library preparation kit (sold separately) provides all the reagents for generating DNA-Seq libraries on the MagicPrep™ NGS system. This kit features a broad input range from 50 to 500 ng of intact genomic DNA, robust enzymatic DNA fragmentation and UDI adaptors for flexible multiplexing. This kit together with the MagicPrep™ NGS system provides a fully automated solution for DNA-Seq library preparation with a 10 minute setup time.
The Revelo mRNA-Seq for MagicPrep™ NGS library preparation kit (sold separately) provides all the reagents for generating stranded mRNA-Seq libraries on the MagicPrep™ NGS system. This kit features a broad input range from 10 to 1000 ng of total RNA and UDI adaptors for flexible multiplexing. This kit together with the MagicPrep™ NGS system provides a fully automated solution for mRNA-Seq library preparation with a 10 minute setup time.
Specificaties
- Gewicht:18,8 kg
- Omschrijving:Instrument Magicprep™ NGS with cable UK; fully automated preparation of 8 next generation sequencing (NGS) libraries
- Seq:03
- Maat:580×250×310 mm