0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
[EN]MASSIVEV PURIFICATION BUFFER SET 1 * 1 EA
WAKF295-96601
:
FUJIFILM WAKO CHEMICALS
[EN]MASSIVEV PURIFICATION BUFFER SET 1 * 1 EA
WAKF295-96601
:
FUJIFILM WAKO CHEMICALS
Frequently Bought Together
VWR®, Microscoopglaasjes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Petrischalen, PS
: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
Pasteurpipetten
: RATIOLAB
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR® Disposable Microcentrifuge Tubes
: VWR Collection
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
VWR®, Centrifugebuizen, metaalvrij, met schroefdop
: VWR Collection
VWR® Standaardlijntransferpipetten
: VWR Collection