Wire Baskets for Test Tubes
140×140×140 mm
Catalog # LINK51108211414
Supplier: LINKER INDUSTRIE TECHN
Jump to:
Product Details & Documents
Wire basket of welded mesh w 8 mm, d 1 mm for test tubes and more. Made up of stainless steel W-no. 1.4301, cube shape, electrochemically polished.
Specifications
- Seq:03
- b×d×h:140×140×140 mm