Wire Baskets for Test Tubes
100×100×100 mm
Catalogus # LINK51108211010
Leverancier: LINKER INDUSTRIE TECHN
Ga naar:
Product Details & Documents
Wire basket of welded mesh w 8 mm, d 1 mm for test tubes and more. Made up of stainless steel W-no. 1.4301, cube shape, electrochemically polished.
Specificaties
- Seq:01
- b×d×h:100×100×100 mm