Orders LinkContactUsLinkComponent
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, light grey,handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×131,0×135,0 mm
Catalogus # BRDY176687
Leverancier:   Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Catalogus # BRDY176687
Leverancier:  Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, light grey,handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×131,0×135,0 mm
Price
1 170,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Easily read wide and small barcodes with the ultra-fast CR2700 Handled barcode scanner. This aggressive, omnidirectional scanner eliminates repositioning and time consuming re-scans of barcodes.
  • Uses B27 batteries with built-in power gauge
  • IP 65 seal rating against dust and liquid ingress
  • CodeShield® housing for disinfectant and chemical wipe down
  • Unequalled performance reading on curved and shiny surfaces
  • USB, Bluetooth®5 Low Energy (for enhanced data security, extended battery life, and less downtime)
Customisable visual, audible and haptic feedback with multiple programmable buttons. Trace any troubles with trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Specificaties
  • Omschrijving:
    CR2700 scanner, light grey,handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×131,0×135,0 mm
  • Seq:
    03
Accessories
Productinformatie
  • Catalogus #
    Voor
    Beschikbaarheid
    Price
  • Workstation lockout writer software app
    Workstation lockout writer software app
    BRDY145445
    BBP®31, BBP®33, BBP®85, i3300
    Log in voor beschikbaarheid
    1 * 1 ST - 833,00 €
Productinformatie
  • Catalogus #
    Voor
    Beschikbaarheid
    Price
  • Brady workstation scan- en printsoftwarepakket
    Brady workstation scan- en printsoftwarepakket
    BRDY149445
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    Log in voor beschikbaarheid
    1 * 1 ST - 833,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Brady workstation CLP/GHS app via download
    Brady workstation CLP/GHS app via download
    BRDY195526
    Log in voor beschikbaarheid
    1 * 1 ST - 369,99 €
Productinformatie
  • Catalogus #
    Voor
    Beschikbaarheid
    Price
  • Brady workstation automation software suite
    Brady workstation automation software suite
    BRDY149187
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    Log in voor beschikbaarheid
    1 * 1 ST - 833,00 €