Cerex® HPSCX SPE Columns
Column Cerex HPSCX, HPSCX, 30 µm, 6 ml, 50 mg
Catalogus # TCAT427-0506C
Leverancier: Tecan
Ga naar:
Product Details & Documents
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
- Ion-exchange
- Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Specificaties
- Partikelgrootte:30 µm
- Capaciteit:6 ml
- Omschrijving:Column Cerex HPSCX
- Seq:02
- Bedgewicht:50 mg
- Fase:HPSCX