Resolvex® Column Temperature Controller
Column cooler 250 CERA, 1,5×4,0×17"
Catalogus # TCAT282-0300
Leverancier: Tecan
Ga naar:
Product Details & Documents
Column temperature controllers for heating and cooling.
- Reliable PID microprocessor controlled thermal circuit stabilizes the operating environment
- Easily-seated thermal blanket encloses the column in horizontal, vertical or slant orientation
- Heater inserts are modular for easy removal and arrangement
Based on a unique, space-saving concept for efficient temperature control of liquid chromatography columns. Column enclosure accepts ³/₈" ext. Ø HPLC columns of any length up to 30 centimeters (15 centimeters in Column Heater 150).
Specificaties
- Omschrijving:Column cooler 250 CERA, 1,5×4,0×17"
- Seq:03