Assistent® Pipette Brushes
Pipette brushes for measuring pipettes 1 - 10 ml, —, —
Catalog # HECH42588010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Brushes for cleaning an assortment of pipette types.
- Galvanised wire handle
- Available with natural or nylon bristles
Documents
TDS Assistent Pipette Brushes.pdf
Datasheet
Specifications
- Ø borstelkop:—
- Omschrijving:Pipette brushes for measuring pipettes 1 - 10 ml
- Seq:04
- Lengte borstelkop:—