[EN]CANISTER. 10 LITER. S60/61. TYPE 8 1 * 1 ST
Catalogus # SCTV108330
Leverancier: S.C.A.T.
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
VWR®, Nitrilhandschoenen
Leverancier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
VWR®, Short Thread Vials ND9
Leverancier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Leverancier: VWR Collection
VWR®, Vacuümfiltratiesystemen, PES
Leverancier: VWR Collection
TLC- en GLP-platen, kiezelgel 60, ongemodifiëerd, glazen platen
Leverancier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
Centrifugebuisjesfilters, Costar® Spin-X®
Leverancier: Corning
VWR®, Snap Cap Vials
Leverancier: VWR Collection
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals