Square HDPE Bottle
500 ml, Square bottle, HDPE
Catalogus # SAMP8165A-500
Leverancier: Sampling Systems
Ga naar:
Product Details & Documents
Square HDPE bottles for use with a range of liquids and powders.
- Different sizes available
- Ribbed sides for easy handling
- Available from stock
- Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.
Specificaties
- Capaciteit:500 ml
- Omschrijving:Square bottle
- Materiaal:HDPE
- Seq:02