LAVA Multirisk Jackets
Multirisk, marineblauw/blauw, L, Regular
Catalog # ALSW1030949-L
Supplier: ALSICO
Jump to:
Product Details & Documents
Comfortable, multirisk jacket in inherent flame retardant fabric.
- Modern fit
- Upstanding rounded collar
- Front closure with hidden studs
- ECO design
- Inside zipped pocket
- 2 large side pockets with flap and hidden press studs
- Long sleeves with elasticated sleeve cuffs
- Piping in accent colour orange
Industrial launderable.
: Oeko-Tex® standard 100; EN 1149-5 2018; IEC 61482 - 2: 2018 - APC1; EN ISO 11612:2015 (A1, B1, C1, D0, E0, F1); EN 11611:2015 - Class 1 A1; EN 13034 +A1.
:
:
:
:
Specifications
- Confectiemaat:L
- Pasvorm van kledingstuk:Regular
- Type:Multirisk
- Kleur:marineblauw/blauw
- Seq:03
- Geslacht:Voor mannen
- Cat. No.:ALSW1030949-L
- Kledingstuk toepassing:Chemical protection|Fire protection|Welding|Industrial|Mechanical protection
- Sleeve or leg length:Long
- Risk area:Flame-retardant|Anti-static
- Sluitingstype:Met drukknopen
- Gewicht:320 g/m²
- Stof:54% modacrylic/44% cotton/2% carbon