Double Ended Spoons
Double ended spoon, 210 mm, 1,6/3 ml, Roestvrij staal
Catalogus # SAMPA104-210
Leverancier: Sampling Systems
Ga naar:
Product Details & Documents
Double ended spoon for taking different sample sizes.
- Made from stainless steel
- Ideal for sampling powders and liquids
- Various sizes available
These double ended spoons are great for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary allowing for flexibility in sampling both powders and liquids.
Specificaties
- Capaciteit:1,6/3 ml
- Omschrijving:Double ended spoon
- Lengte:210 mm
- Materiaal:Roestvrij staal
- Seq:01