Xyleen ≥98%, TECHNICAL
950 L
Leverancier: VWR Chemicals
Ga naar:
Product Details & Documents
Synoniemen: Dimethylbenzene
- CAS-nummer:1330-20-7
- Formule:C₆H₄(CH₃)₂
- MDL-nummer:MFCD00077264
- Molecular Weight:106.17 g/mol
- Merck index:13,10136
- Kokend Pt:140 °C (1013 hPa)
- Smelten Pt:–34 °C
- Dichtheid:0,86 g/cm³ (20 °C)
- Flits Pt:25 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:215-535-7
- UN:1307
- ADR:3,III
- BEREIKEN:01-2119488216-32
Documents
Certificaten
Zoeken op lotnummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC28973.294_SPEC_EN.pdf P20_VWRC28973.328_SPEC_EN.pdf P20_VWRC28973.363_SPEC_EN.pdf P20_VWRC28973.465_SPEC_EN.pdf spec_VWRC28973.294
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specificaties
Specificatie testresultaten
- Assay:Min. 98%
Specificaties
- VPE:950 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



