Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Qualitative Test Papers, Cobalt
Qualitative Test Papers, Cobalt
QUANTOFIX Phosphate 10, 0 • 0.5 • 2.0 • 5.0 • 10.0 - mg/L - PO₄³⁻, Molybdenum blue reaction, 40×40×120 mm (package), 50 test(s)
Catalogus # MANA91356
Leverancier:   MACHEREY-NAGEL
Qualitative Test Papers, Cobalt
Catalogus # MANA91356
Leverancier:  MACHEREY-NAGEL
CAS-nummer:  
Qualitative Test Papers, Cobalt
Qualitative Test Papers, Cobalt
QUANTOFIX Phosphate 10, 0 • 0.5 • 2.0 • 5.0 • 10.0 - mg/L - PO₄³⁻, Molybdenum blue reaction, 40×40×120 mm (package), 50 test(s)
CAS-nummer:  
Price
29,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
QUANTOFIX® test strips meet all requirements of a modern rapid test. The colour of the reactive pad changes depending on the concentration of an analyte in the sample. The evaluation is usually carried out visually by comparison of the reaction colour with a multi-stage colour scale. The test strips are intended for single use, maintenance or calibration are not required.
  • Results within seconds: Just dip, wait and read against multi-stage colour scale Always ready to use: No accessories necessary for evaluation Safe results: Desiccant in the stopper for optimal protection of the strips against humidity
These test strips are ideal to test levels of phosphate in solutions. The easy 'dip and read' procedure provides realiable results within 90 seconds.
Specificaties
Specificaties
  • Chemische methode:
    Molybdenum blue reaction
  • Omschrijving:
    QUANTOFIX Phosphate 10
  • Afmetingen:
    40×40×120 mm (package)
  • Tests per kit:
    50 test(s)
  • Meetbereik:
    0 • 0.5 • 2.0 • 5.0 • 10.0 - mg/L - PO₄³⁻
  • Platform:
    Semi-quantitative test strips
  • Method:
    Phosphate - Phosphomolybdenum blue (yellow-green → green-blue)
  • Gewicht:
    35 g
  • Cat. No.:
    MANA91356
  • Instrument:
    No
  • Parameter:
    All reagents included
  • Bestanddelen:
    50 test strips in a tube, instructions for use
  • Conformiteit:
    No hazardous material
  • Houdbaarheid (in maanden):
    30 (from production)
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.