Acetonitril ≥99%, TECHNICAL
950 L
Leverancier: VWR Chemicals
Ga naar:
Product Details & Documents
Synoniemen: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS-nummer:75-05-8
- Formule:H₃CCN
- MDL-nummer:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck index:13,00071
- Kokend Pt:81,6 °C (1013 hPa)
- Smelten Pt:–45,7 °C
- Dichtheid:0,782 g/cm³ (20 °C)
- Flits Pt:2 °C (gesloten beker)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-835-2
- UN:1648
- ADR:3,II
- BEREIKEN:01-2119471307-38
Documents
Certificaten
Zoeken op lotnummer1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 30L Metal drum for liquids UN X (ZPACK12124125)
Packaging Information
P20_VWRC20070.291_SPEC_EN.pdf P20_VWRC20070.325_SPEC_EN.pdf P20_VWRC20070.462_SPEC_EN.pdf P20_VWRC20070.553_SPEC_EN.pdf P20_VWRC20070.950_SPEC_EN.pdf
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specificaties
Specificatie testresultaten
- Assay:Min. 99 %
- Refractive index (20°C):1.343 - 1.345
Specificaties
- VPE:950 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



