Orders LinkContactUsLinkComponent
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, dark grey, handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×132×134,6 mm
Catalog # BRDY176516
Supplier:   Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Catalog # BRDY176516
Supplier:  Brady
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
Handheld Wireless Handle Barcode Scanner; Bluetooth, 1D, 2D, QR Code, CR2700
CR2700 scanner, dark grey, handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×132×134,6 mm
Price
1 170,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Easily read wide and small barcodes with the ultra-fast CR2700 Handled barcode scanner. This aggressive, omnidirectional scanner eliminates repositioning and time consuming re-scans of barcodes.
  • Uses B27 batteries with built-in power gauge
  • IP 65 seal rating against dust and liquid ingress
  • CodeShield® housing for disinfectant and chemical wipe down
  • Unequalled performance reading on curved and shiny surfaces
  • USB, Bluetooth®5 Low Energy (for enhanced data security, extended battery life, and less downtime)
Customisable visual, audible and haptic feedback with multiple programmable buttons. Trace any troubles with trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Spécifications
  • Omschrijving:
    CR2700 scanner, dark grey, handled version, kit with B27 battery, inductive charging station with desktop mount, and 1,8 m USB charging cable, W×D×H: 53,3×132×134,6 mm
  • Seq:
    01
Accessories
Informations sur le produit
  • Catalog #
    Omschrijving
    Voor
    Availability
    Price
  • BRDY145445
    Workstation lockout writer software app
    BBP®31, BBP®33, BBP®85, i3300
    Connexion Pour la disponibilité
    1 * 1 ST - 833,00 €
Informations sur le produit
  • Catalog #
    Omschrijving
    Voor
    Availability
    Price
  • BRDY149445
    Brady workstation scan- en printsoftwarepakket
    BBP11, BBP12, BBP31, BBP33, BBP35, BBP37, BBP81, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Colour and Cut, GlobalMark 2 Multicolour, i3300, i5100, i7100, i7500, IDXPERT, IP Series, J2000, J4000, J5000, LABXPERT, M410, M510, M610, M611, M710, PR Plus, S3100, THT 3" Core Printers, TLS 2200
    Connexion Pour la disponibilité
    1 * 1 ST - 833,00 €
Informations sur le produit
  • Catalog #
    Omschrijving
    Availability
    Price
  • BRDY195526
    Brady workstation CLP/GHS app via download
    Connexion Pour la disponibilité
    1 * 1 ST - 369,99 €
Informations sur le produit
  • Catalog #
    Omschrijving
    Voor
    Availability
    Price
  • BRDY149187
    Brady workstation automation software suite
    BBP11, BBP12, BBP30, BBP33, BBP35, BBP37, BBP85, BMP41, BMP51, BMP53, BMP61, BMP71, GlobalMark 2 Multicolor, i3300, i5100, i7100, IP Series, J2000, J4000, J5000, M610, M611, M710, PR Plus, S3000, THT 76 mm Core Printers, Wraptor, Wraptor A6500
    Connexion Pour la disponibilité
    1 * 1 ST - 833,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.