Ethanol, GPR RECTAPUR®
950 L
Leverancier: VWR Chemicals
Ga naar:
Product Details & Documents
Caution: WAARSCHUWING: Deze alcohol is gereguleerd. Bel ons.
Synoniemen: Ethyl alcohol
- CAS-nummer:64-17-5
- Formule:C₂H₅OH
- MDL-nummer:MFCD00003568
- Molecular Weight:46.07 g/mol
- Kokend Pt:78,3 °C (1013 hPa)
- Smelten Pt:–117 °C
- Dichtheid:0,7895 g/cm³ (20 °C)
- Flits Pt:12 °C (gesloten beker)
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- BEREIKEN:01-2119457610-43
Documents
Certificaten
Zoeken op lotnummer1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
20824_ETHANOL 96 % VOL GPR RECTAPUR_BSE_REV0
BSE/TSE Certificates
P20_VWRC20824.296_SPEC_EN.pdf P20_VWRC20824.321_SPEC_EN.pdf P20_VWRC20824.365_SPEC_EN.pdf P20_VWRC20824.467_SPEC_EN.pdf P20_VWRC20824.558_SPEC_EN.pdf
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specificaties
Specificatie testresultaten
- Assay (on anhydrous substance):Min. 99.8 %
- Assay (V/V):96.0 - 96.3 %
- Acidity:Max. 0.0002 meq/g
- Aldehydes (as CH3CHO):Max. 15 ppm
- Evaporation residue:Max. 20 ppm
Specificaties
- VPE:950 L
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



