Order Entry
Belgium
Orders LinkContactUsLinkComponent
ethanedithioamide 98%

ethanedithioamide 98%

Bottle
Catalogus # APOSOR24680-250MG
Irriterend
Leverancier:   Apollo Scientific
ethanedithioamide 98%
Catalogus # APOSOR24680-250MG
Leverancier:  Apollo Scientific
CAS-nummer:  79-40-3
ethanedithioamide 98%

Bottle

CAS-nummer:  79-40-3
Price
21,80 €
Log in voor beschikbaarheid
1 * 250 mg
Product Details & Documents

Usage: Organic building block/reagent.

Synoniemen:  Ethanebis(thioamide), Rubeanic acid
  • CAS-nummer:79-40-3
  • Formule:C₂H₄N₂S₂
  • MDL-nummer:MFCD00004941
  • Molecular Weight:120,19655 g/mol
  • EINECS:201-203-9
Specificaties

Specificaties

  • Verpakking:
    Bottle

Specificaties