Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Poly(vinylidene fluoride)

Bottle, 5 g
Catalogus # APOSPC37872-5G
Irriterend
Leverancier:   Apollo Scientific
Poly(vinylidene fluoride)
Catalogus # APOSPC37872-5G
Leverancier:  Apollo Scientific
CAS-nummer:  24937-79-9
Poly(vinylidene fluoride)

Bottle, 5 g

CAS-nummer:  24937-79-9
Price
29,00 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
  • CAS-nummer:24937-79-9
  • Formule:(-CH₂CF₂-)n
  • MDL-nummer:MFCD00084470
  • Smelten Pt:155…165 °C
  • Dichtheid:1,74 g/cm³ (25 °C)
  • Temperatuur bij opslag:Kamertemperatuur
Specificaties

Specificaties

  • Verpakking:
    Bottle
  • VPE:
    5 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.