Masterflex® Nylon Clamp for Sanitary Fittings, 2"; 10/PK
Catalogus # MFLX12805-05
Leverancier: Avantor Fluid Handling
Over dit artikel
Collared clamp helps prevent unintentional opening.
- All fittings are compatible with Tri-Clamp® fittings
- Excellent chemical resistance; low leachable characteristics
- Omschrijving:Nylon Clamp for Sanitary Fittings, 2"; 10/PK
- Materiaal:Nylon
- Seq:03
- Environmentally Preferable:
- Sanitary clamp size (port 1):2"
- Sterilisatie:Autoclave, Gamma
- VPE:10
- Klemtype:Sanitary
Frequently Bought Together