GC septa
BTO premium high-temperature centre guide GC septa, 11,5 mm, 400 °C
Catalogus # SGEA298777
Leverancier: Trajan Scientific and Medical
Ga naar:
Product Details & Documents
Specificaties
- Ø:11,5 mm
- Max. temp.:400 °C
- Omschrijving:BTO premium high-temperature centre guide GC septa
- Seq:15