Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
PTFE GC Ferrules

PTFE GC Ferrules

GC ferrule 3,17 mm, PTFE, 1,58 mm
Catalogus # SGEA214210
PTFE GC Ferrules
Catalogus # SGEA214210
PTFE GC Ferrules

GC ferrule 3,17 mm, PTFE, 1,58 mm

Price
16,70 €
Log in voor beschikbaarheid
1 * 10 ST
Product Details & Documents

These GC ferrules are made from 100% PTFE. They are soft, completely inert, and have very low friction.

PTFE GC ferrules are hydrophobic and commonly used in liquid applications. They are rated upto upper temperature limit of 250 °C.

Specificaties
  • Ø inw.:
    1,58 mm
  • Omschrijving:
    GC ferrule 3,17 mm
  • Materiaal:
    PTFE
  • Seq:
    11