Inbed-ringen
blauw
Catalogus # KART0291904
Leverancier: KARTELL
Ga naar:
Product Details & Documents
Embedding rings made from ABS.
- Dimensions: 41×28×11 mm
Available in the colours white, blue, yellow, green and pink.
Specificaties
- Kleur:blauw
Embedding rings made from ABS.
Available in the colours white, blue, yellow, green and pink.