Polka Dot Plus Gloves
M, wit/blauw
Catalogus # PRTWA113WBRM
Ga naar:
Discontinued
Product Details & Documents
PVC dots on both sides extend the life of this glove. Perfect for warehousing and distribution, these ambidextrous gloves can be worn on either hand.
- PVC dotted palm for enhanced grip
- Ambidextrous design suitable for left- and right-handed users
- Seamless 13 gauge liner
- Performs well in dry conditions
Low linting construction for minimal contamination, with a breathable seamless liner.
Colour: White/blue
Certifications: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 1142X. CE certified.
Documents
TDS A113 - Polka Dot Plus Gloves
Datasheet
Specificaties
- Handschoenmaat:M
- Kleur:wit/blauw
- Seq:02
- Gepoederd/poedervrij:poedervrij
- Cat. No.:PRTWA113WBRM
- Voor eenmalig gebruik/herbruikbaar:Herbruikbaar
- Latex/latexvrij:Latexvrij
- Ambidexter/handspecifiek:Ambidexter
- Materiaal:Polyester
- Sterility:Non sterile