Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Valproic Acid, Reference Standards

Valproic Acid, Reference Standards

Valproic Acid Related Compound K, USP reference standard, Ampule, 25 mg
Catalogus # 1708650.
Leverancier:   USP
Valproic Acid, Reference Standards
Catalogus # 1708650.
Leverancier:  USP
CAS-nummer:  
Valproic Acid, Reference Standards

Valproic Acid Related Compound K, USP reference standard, Ampule, 25 mg

CAS-nummer:  
Price
1 110,00 €
Log in voor beschikbaarheid
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Ampule
  • VPE:
    25 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Valproic Acid Related Compound K
  • Cat. No.:
    1708650.
  • Omschrijving:
    Valproic Acid Related Compound K
  • Synonym(s):
    2-Ethyl-2-methylpentanoic acid
  • CAS No.:
    5343-52-2

Specificaties