Order Entry
Belgium
Orders LinkContactUsLinkComponent
Etoposide, Reference Standards

Etoposide, Reference Standards

Etoposide Phosphate, USP reference standard, Vial, 200 mg
Catalogus # 1268819.
Leverancier:   USP
Etoposide, Reference Standards
Catalogus # 1268819.
Leverancier:  USP
CAS-nummer:  
Etoposide, Reference Standards

Etoposide Phosphate, USP reference standard, Vial, 200 mg

CAS-nummer:  
Price
555,00 €
Log in voor beschikbaarheid
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certificeringen: US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Verpakking:
    Vial
  • VPE:
    200 mg
  • Standaard:
    USP reference standard
  • Omschrijving:
    Etoposide Phosphate
  • UN No.:
    1325
  • Cat. No.:
    1268819.
  • Omschrijving:
    Etoposide Phosphate
  • CAS No.:
    117091-64-2

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.