0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
[EN]12 MM TRANSWELL 0.4 UM PORE INSERTS 1 * 48 ST
STMC38023
:
STEMCELL Technologies
[EN]12 MM TRANSWELL 0.4 UM PORE INSERTS 1 * 48 ST
STMC38023
:
STEMCELL Technologies
Frequently Bought Together
VWR® Standaardlijntransferpipetten
: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Transferpipetten
: Brand
VWR® Premium Line, Serologische pipetten
: VWR Collection
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
VWR®, Centrifugebuizen, Vrijstaand, met Platte Schroefdoppen
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR®, Centrifugeerbuisjes, conische bodem
: VWR Collection
VWR®, Centrifugebuizen, metaalvrij, met schroefdop
: VWR Collection