SWIN Tips, small (600 μg, i.d. 0.6 mm), glass, 1000 pcs Pack of assembled (capillary and piston,combined in common container) Pick & Place glass tips for SWILE (GDU-S). For an approximate dispense range 200-1000 μg. 1 * 1.000 ST
Catalog # MSPPB0920.1096
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together