Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Inserts for U-D master manual samplers

Inserts for U-D master manual samplers

U-D Master insert split, Roestvrij staal 316L, 0,3 ml
Catalogus # SAMP1230T-03S
Leverancier:   Sampling Systems
Inserts for U-D master manual samplers
Catalogus # SAMP1230T-03S
Leverancier:  Sampling Systems
Inserts for U-D master manual samplers
Inserts for U-D master manual samplers

U-D Master insert split, Roestvrij staal 316L, 0,3 ml

Price
549,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Stainless steel inserts for U-D Master manual samplers.

  • Made from 316 stainless steel

These inserts are ideal for helping to prevent sampling error. By simply changing the sample inserts, different volumes can be accurately sampled.

Specificaties
  • Volume:
    0,3 ml
  • Omschrijving:
    U-D Master insert split
  • Materiaal:
    Roestvrij staal 316L
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.