Objectives for Rebel Microscopes
Plan fluorite POL oil NA 1,3, 100×, 0,2 mm
Catalogus # ECHLJMAG1010
Leverancier: DISCOVER ECHO
Ga naar:
Product Details & Documents
Documents
Specificaties
- Vergroting:100×
- Omschrijving:Plan fluorite POL oil NA 1,3
- Seq:04
- Werkafstand:0,2 mm