100 ML INFUSION VIAL MADE OF TYPE 1 CLEAR GLASS WITH CRIMP NECK S 32 MM. PHARMACEUTICAL GRADE MOULDED GLASS. SGD MOLD REFERENCE 3435 8. TYPE: ISO INFUSION VIAL. DIMENSIONS (OD X H) 49 MM X 104 MM. SGD MOLD REFERENCE 34358. 1 * 4.160 Flacon
Catalogus # MSPP1000125
Leverancier: MARKET SOURCE PART PROCESS
Frequently Bought Together