Masterflex® Fitting, Nylon, Straight, Male Luer to Hose Barb Adapter, 5/32" ID; 25/PK (use with Separate Lock Ring)
Catalogus # MFLX45505-46
Leverancier: Avantor Fluid Handling
CE certificate
Avantor exclusive
Over dit artikel
Verzeker uw procesintegriteit met volledige materiaaltraceerbaarheid.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Male Luer x 5/32" ID (use with separate lock ring)
- Ideal for use with flexible tubing
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Weerhaakmaat/buis inw.Ø (poort 1):⁵⁄₃₂"
- Verbindingstype:Male luer to barb
- Flow pattern:Straight
- Omschrijving:Male luer×⁵⁄₃₂" hose barb (for separate lock ring), nylon, 25/pk
- Seq:18
- Environmentally Preferable:
- Model:45505-46
- Connection type (port 1):Male luer slip
- No. of ports:2
- Garantie:3 Months
- VPE:25
- Fitting material:Nylon
Frequently Bought Together