LITHIUM HEXAFLUOROPHOSPHATE, 99+% 1 * 5 g
Catalogus # STRE93-0325-5G
Leverancier: Strem Chemicals, Inc.
LITHIUM HEXAFLUOROPHOSPHATE, 99+% 1 * 5 g
Catalogus # STRE93-0325-5G
Leverancier: Strem Chemicals, Inc.
Frequently Bought Together
Ammonium sulphate ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
Natriumnitriet ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Potassium chloride, EMSURE® for analysis, Supelco®
Leverancier: Merck
Natriumchloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Leverancier: Merck
Natriumchloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Natriumchloride ≥98%, TECHNICAL
Leverancier: VWR Chemicals
Natriumsulfaat, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Leverancier: VWR Chemicals
EDTA disodium salt dihydrate 99.0-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Leverancier: VWR Chemicals
Calciumchloride 90-98%, TECHNICAL
Leverancier: VWR Chemicals
Magnesium oxide light 98.0-100.5%, AnalaR NORMAPUR® analytical reagent
Leverancier: VWR Chemicals