LITHIUM HEXAFLUOROPHOSPHATE, 99+% 1 * 25 g
Catalogus # STRE93-0325-25G
Leverancier: Strem Chemicals, Inc.
LITHIUM HEXAFLUOROPHOSPHATE, 99+% 1 * 25 g
Catalogus # STRE93-0325-25G
Leverancier: Strem Chemicals, Inc.
Frequently Bought Together
LITHIUM BROMIDE (99.95%-LI) (0.2% H2O) 1 * 50 g
Leverancier: Strem Chemicals, Inc.
ZIRCONIUM ROD (99.8%) 1 * 5 cm
Leverancier: Strem Chemicals, Inc.
Azijnzuur glacial 99-100%, BAKER ANALYZED® ACS, J.T.Baker®
Leverancier: Avantor
RUBIDIUM (99.9+%) (PRESCORED AMPOULE) 1 * 1 g
Leverancier: Strem Chemicals, Inc.
NIOBIUM(V) OXIDE (99.9%-NB) 1 * 250 g
Leverancier: Strem Chemicals, Inc.
ANTIMONY(III) OXIDE, ELEC. GR. (99.999%- 1 * 25 g
Leverancier: Strem Chemicals, Inc.
SILVER NANOPARTICLES (40NM, 0.02MG/ML IN 1 * 20 mL
Leverancier: Strem Chemicals, Inc.
SODIUM L-GLUTAMATE 1-HYDRATE (USP-NF) PU 1 * 5 kg
Leverancier: PanReac AppliChem
Koper(II)sulfaat pentahydraat 98.0-102.0% ACS
Leverancier: Thermo Fisher Scientific
Iron(III) chloride hexahydrate 99+% for analysis
Leverancier: Thermo Fisher Scientific
COBALT(II) PERCHLORATE HEXAHYDRATE, 98+% 1 * 50 g
Leverancier: Strem Chemicals, Inc.
di-Potassium hydrogen phosphate ≥99.0% analytical reagent
Leverancier: VWR Chemicals
MAGNESIUM SULFATE 7-HYDRATE FOR ANALYSIS 1 * 1.000 g
Leverancier: PanReac AppliChem
TUNGSTEN(VI) OXIDE, SINTERED LUMPS (99.9 1 * 10 g
Leverancier: Strem Chemicals, Inc.