LITHIUM HEXAFLUOROPHOSPHATE (99.9+%-LI) 1 * 10 g
Catalogus # STRE03-0325-10G
Leverancier: Strem Chemicals, Inc.
LITHIUM HEXAFLUOROPHOSPHATE (99.9+%-LI) 1 * 10 g
Catalogus # STRE03-0325-10G
Leverancier: Strem Chemicals, Inc.
Frequently Bought Together
Cobalt(II) sulphate heptahydrate, EMSURE® for analysis, Supelco®
Leverancier: Merck
di-natriumwaterstoffosfaat-heptahydraat geschikt voor biofarmaceutische productie EMPROVE bio DAC,USP 1 * 5 kg
Leverancier: MERCK PRODUCTION CHEMICALS
α-Ionone, Sigma-Aldrich®
Leverancier: Merck
Aluminium nitrate nonahydrate, Supelco®
Leverancier: Merck
Natriumdiwaterstoffosfaat dihydraat, BAKER, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
RUTHENIUM(III) NITROSYL CHLORIDE MONOHYD 1 * 5 g
Leverancier: Strem Chemicals, Inc.
Zinc stearate
Leverancier: Thermo Fisher Scientific
Ammoniumchloride 99.5%, extra pure
Leverancier: Thermo Fisher Scientific
HAFNIUM(IV) CHLORIDE, SUBLIMED GRADE (99 1 * 50 g
Leverancier: Strem Chemicals, Inc.
GOLD NANOPARTICLES (50NM DIAMETER, 1 OD, 1 * 25 mL
Leverancier: Strem Chemicals, Inc.
Disodium hydrogen phosphate heptahydrate 98.0-102.0% ACS
Leverancier: Thermo Fisher Scientific
Disodium hydrogen phosphate dihydrate 99+% for HPLC
Leverancier: Thermo Fisher Scientific
ZINC OXIDE (99.9995%-ZN) PURATREM 1 * 500 g
Leverancier: Strem Chemicals, Inc.
MAGNESIUM NITRATE HEXAHYDRATE, 99% (ACS) 1 * 1 kg
Leverancier: Strem Chemicals, Inc.
BORON POWDER, CRYSTALLINE (99.99%) 1 * 1 g
Leverancier: Strem Chemicals, Inc.
INDIUM (III) NITRATE PENTAHYDRATE (99.99 1 * 50 g
Leverancier: Strem Chemicals, Inc.
Lanthanum(III) oxide ≥99.5%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Potassium bromide 99.5-100.5%, Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Leverancier: Honeywell Chemicals