Orders LinkContactUsLinkComponent
Slangen
Slangen
6,00 mm, 2,00 mm, 10,00 mm
Catalogus # DENE3500610
Leverancier:   DEUTSCH NEUMANN
Slangen
Catalogus # DENE3500610
Leverancier:  DEUTSCH NEUMANN
Slangen
Slangen
6,00 mm, 2,00 mm, 10,00 mm
Price
265,00 €
Log in voor beschikbaarheid
1 * 100 m
Product Details & Documents
PVC, transparant
  • High flexibility, good chemical resistance
  • Smooth both inside and out, excellent resistance to abrasion
  • Good value, large size rolls
Universal tubing for a wide range of applications. The preferred option where the flow of liquids has to be observed. Not suitable for operation under vacuum or pressure.
Temperature resistance: −20 to +60 °C
Hardness (Shore durometer A): 77
Specificaties
  • Buis ext.Ø:
    10,00 mm
  • Buis int.Ø:
    6,00 mm
  • Wanddikte:
    2,00 mm
  • Seq:
    30