Parker Hannifin Compression Union Tee Adapter, SS, 1/8" OD (2) x 1/8" NPT(M); Each
Catalogus # MFLX31406-85
Leverancier: Avantor Fluid Handling
CE certificate
Over dit artikel
Get a secure seal at higher pressures.
- Quick and easy assembly without tools
Compression and threaded fittings offer a more permanent installation. Choose the size and material to suit your application needs.
- Verbindingstype:Compression to Thread
- Flow pattern:T
- Omschrijving:Compression Union Tee Adapter, SS, 1/8" OD (2)×1/8" NPT(M); Each
- Environmentally Preferable:
- Sterilisatie:Autoclave, ethylene oxide (ETO), or gamma
- Garantie:LTD life time
- VPE:1
- Fitting material:Stainless Steel 316
Frequently Bought Together